missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific™

Catalog Number 10561944
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
613239-76-2
C13H9ClF3N
271.67
MFCD09817479
JTXHHQDYEHXWPC-UHFFFAOYSA-N
2-4-chloromethyl phenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl pyridin-2-yl benzyl chloride, 2-4-chloromethyl-phenyl 5-trifluoromethyl-pyridine, 2-4-chloromethyl-phenyl-5-trifluoromethyl-pyridine
22280067
2-[4-(chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(CCl)C=C1
This item is not returnable. View return policy
613239-76-2
C13H9ClF3N
271.67
MFCD09817479
JTXHHQDYEHXWPC-UHFFFAOYSA-N
2-4-chloromethyl phenyl-5-trifluoromethyl pyridine, 4-5-trifluoromethyl pyridin-2-yl benzyl chloride, 2-4-chloromethyl-phenyl 5-trifluoromethyl-pyridine, 2-4-chloromethyl-phenyl-5-trifluoromethyl-pyridine
22280067
2-[4-(chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(CCl)C=C1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 271.67
Percent Purity 95%
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'