missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Chloromethyl)phenyl]-1,3-thiazole, 97%, Maybridge™

£61.28 - £158.00

Chemical Identifiers

CAS 906352-61-2
Molecular Formula C10H8ClNS
Molecular Weight (g/mol) 209.691
MDL Number MFCD09064951
InChI Key BFAWMDHRKPPXSO-UHFFFAOYSA-N
Synonym 2-4-chloromethyl phenyl-1,3-thiazole, 2-4-chloromethyl phenyl thiazole
PubChem CID 24229488
IUPAC Name 2-[4-(chloromethyl)phenyl]-1,3-thiazole
SMILES C1=CC(=CC=C1CCl)C2=NC=CS2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10396804
SDS View Documents
Maybridge
CC40224CB
250mg Amber Glass Bottle
£61.28
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 30-06-2021
Please to purchase this item. Need a web account? Register with us today!
10783964
SDS View Documents
Maybridge
CC40224DA
1g Amber Glass Bottle
£158.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

906352-61-2
209.691
BFAWMDHRKPPXSO-UHFFFAOYSA-N
24229488
C1=CC(=CC=C1CCl)C2=NC=CS2
C10H8ClNS
MFCD09064951
2-4-chloromethyl phenyl-1,3-thiazole, 2-4-chloromethyl phenyl thiazole
2-[4-(chloromethyl)phenyl]-1,3-thiazole

Specifications

2-[4-(Chloromethyl)phenyl]-1, 3-thiazole
250mg
96.5
97%
MFCD09064951
BFAWMDHRKPPXSO-UHFFFAOYSA-N
2-[4-(chloromethyl)phenyl]-1,3-thiazole
24229488
97%
Amber Glass Bottle
906352-61-2
100.0
C10H8ClNS
2-4-chloromethyl phenyl-1,3-thiazole, 2-4-chloromethyl phenyl thiazole
C1=CC(=CC=C1CCl)C2=NC=CS2
209.691
209.7
SDS
Product Certifications