missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62150.03

Code : NEW

Additional Details : CAS Number : 69570-83-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407096

  • £28.09 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromophenyl)thiazolidine-4-carboxylic acid
69570-83-8
288.16
MFCD01067251

Specifications

2-(4-Bromophenyl)thiazolidine-4-carboxylic acid
175° to 177°C
Solid
288.16
69570-83-8
1g
97%
MFCD01067251
Videos
SDS