missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)ethylamine, 98%

Brand:  Alfa Aesar™ H63706.03

Code : NEW

Additional Details : CAS Number : 73918-56-6 Weight : 0.00100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15430539

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Applications
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-(4-Bromophenyl)ethylamine
200.08
UN2735
73918-56-6
MFCD00008189
4-Bromophenethylamine

Specifications

2-(4-Bromophenyl)ethylamine
1.5740
63° to 72°C (0.2mmHg)
Liquid
Colorless
MFCD00008189
4-Bromophenethylamine
73918-56-6
1.290g/mL
1g
98%
200.08
UN2735
Videos
SDS