missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-5-methylbenzimidazole, 95%, Thermo Scientific™

Catalog Number 15471059
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
496807-37-5
C14H11BrN2
287.16
MFCD06312161
LWVJBMHQRDXWGN-UHFFFAOYSA-N
2-4-bromophenyl-5-methyl-1h-benzimidazole, 2-4-bromophenyl-6-methyl-1h-benzimidazole, 2-4-bromophenyl-5-methylbenzimidazole, 2-4-bromophenyl-6-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-6-methyl-1h-benzo d imidazole, 2-4-bromophenyl-5-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-5-methyl-3h-1,3-benzodiazole
924735
2-(4-bromophenyl)-6-methyl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=C(C=C3)Br
This item is not returnable. View return policy
496807-37-5
C14H11BrN2
287.16
MFCD06312161
LWVJBMHQRDXWGN-UHFFFAOYSA-N
2-4-bromophenyl-5-methyl-1h-benzimidazole, 2-4-bromophenyl-6-methyl-1h-benzimidazole, 2-4-bromophenyl-5-methylbenzimidazole, 2-4-bromophenyl-6-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-6-methyl-1h-benzo d imidazole, 2-4-bromophenyl-5-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-5-methyl-3h-1,3-benzodiazole
924735
2-(4-bromophenyl)-6-methyl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=C(C=C3)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 287.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)-5-methylbenzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'