missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-5-fluoroindole, 97%, Thermo Scientific™

Catalog Number 15459588
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
885266-74-0
C14H9BrFN
290.135
MFCD05224214
XAKLXLIHCVKNII-UHFFFAOYSA-N
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
24720950
2-(4-bromophenyl)-5-fluoro-1H-indole
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br
This item is not returnable. View return policy
885266-74-0
C14H9BrFN
290.135
MFCD05224214
XAKLXLIHCVKNII-UHFFFAOYSA-N
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
24720950
2-(4-bromophenyl)-5-fluoro-1H-indole
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 290.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)-5-fluoroindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'