missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)-1-phenylbenzimidazole, 97%

Brand:  Alfa Aesar™ H64880.03

Code : NEW

Additional Details : CAS Number : 2620-76-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412279

  • £38.20 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromophenyl)-1-phenylbenzimidazole
349.23
617138
2620-76-0
MFCD09261342

Specifications

2-(4-Bromophenyl)-1-phenylbenzimidazole
162° to 166°C
Powder
Off-white
MFCD09261342
2620-76-0
1g
97%
349.23
617138
Videos
SDS