missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-1,3-dioxolane, 98+%, Thermo Scientific Chemicals

Catalog Number 10677084
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10602-01-4
C9H9BrO2
229.07
MFCD01318950
ZYIMHOWVWWHLDN-UHFFFAOYSA-N
2-4-bromophenyl-1,3-dioxolane, 1-bromo-4-1,3-dioxolan-2-yl benzene, 4-bromobenzaldehyde ethylene acetal, 1,3-dioxolane,2-4-bromophenyl, 2-4'-bromophenyl-1,3-dioxolane, 1,3-dioxolane, 2-4-bromophenyl, bromophenyldioxolane, acmc-2098ib, 2-4-bromophenyl dioxolane, 4-bromobenzaldehyde ethylene ketal
2773842
2-(4-bromophenyl)-1,3-dioxolane
C1COC(O1)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
10602-01-4
C9H9BrO2
229.07
MFCD01318950
ZYIMHOWVWWHLDN-UHFFFAOYSA-N
2-4-bromophenyl-1,3-dioxolane, 1-bromo-4-1,3-dioxolan-2-yl benzene, 4-bromobenzaldehyde ethylene acetal, 1,3-dioxolane,2-4-bromophenyl, 2-4'-bromophenyl-1,3-dioxolane, 1,3-dioxolane, 2-4-bromophenyl, bromophenyldioxolane, acmc-2098ib, 2-4-bromophenyl dioxolane, 4-bromobenzaldehyde ethylene ketal
2773842
2-(4-bromophenyl)-1,3-dioxolane
C1COC(O1)C2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5630 to 1.5650 (20°C, 589nm)
Quantity 1 g
Formula Weight 229.07
Percent Purity 98+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-(4-Bromophenyl)-1, 3-dioxolane
Show More Show Less
compliance-icons
Product Identifier
  • 2-(4-Bromophenyl)-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'