missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)pyridine, ≥95%, Thermo Scientific™

Catalog Number p-3174826
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4783-82-8
C11H8BrNO
250.10
MFCD08435900
FOKJQGKEVMZAJE-UHFFFAOYSA-N
2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
23201935
2-(4-bromophenoxy)pyridine
BrC1=CC=C(OC2=CC=CC=N2)C=C1
This item is not returnable. View return policy
4783-82-8
C11H8BrNO
250.10
MFCD08435900
FOKJQGKEVMZAJE-UHFFFAOYSA-N
2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
23201935
2-(4-bromophenoxy)pyridine
BrC1=CC=C(OC2=CC=CC=N2)C=C1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.09
Percent Purity 95%
Chemical Name or Material 2-(4-Bromophenoxy)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'