missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromobenzyl)thiophene, 97%, Thermo Scientific™

Catalog Number p-3880683
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
118150-25-7
C11H9BrS
253.157
MFCD11841076
GGYCIKURJBHUER-UHFFFAOYSA-N
2-4-bromobenzyl thiophene, 2-4-bromophenyl methyl thiophene
15322628
2-[(4-bromophenyl)methyl]thiophene
C1=CSC(=C1)CC2=CC=C(C=C2)Br
This item is not returnable. View return policy
118150-25-7
C11H9BrS
253.157
MFCD11841076
GGYCIKURJBHUER-UHFFFAOYSA-N
2-4-bromobenzyl thiophene, 2-4-bromophenyl methyl thiophene
15322628
2-[(4-bromophenyl)methyl]thiophene
C1=CSC(=C1)CC2=CC=C(C=C2)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-(4-Bromobenzyl)thiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'