missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromo-2-thienyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62115.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407056

  • £331.78 / 5g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromo-2-thienyl)thiazolidine-4-carboxylic acid
294.19
MFCD08556860

Specifications

2-(4-Bromo-2-thienyl)thiazolidine-4-carboxylic acid
97%
MFCD08556860
Solid
294.19
5g
Videos
SDS