missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromo-2-thienyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64200.06

Code : NEW

Additional Details : CAS Number : 1025843-75-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411309

  • £597.84 / 5g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromo-2-thienyl)benzimidazole
1025843-75-7
279.15
MFCD26967922

Specifications

2-(4-Bromo-2-thienyl)benzimidazole
Solid
279.15
5g
1025843-75-7
95%
MFCD26967922
Videos
SDS