missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole, 95%

Brand:  Alfa Aesar™ H64447.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471659

  • £119.78 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole
313.6
MFCD26967923

Specifications

2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole
95%
MFCD26967923
Solid
313.6
1g
Videos
SDS