missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole, 95%

Brand:  Alfa Aesar™ H64447.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481659

  • £597.84 / 5g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole
313.6
MFCD26967923

Specifications

2-(4-Bromo-2-thienyl)-5-chlorobenzimidazole
95%
MFCD26967923
Solid
313.6
5g
Videos
SDS