missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzyl bromide, 95%, Thermo Scientific Chemicals

Catalog Number p-4228386
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
140690-56-8
C9H5BrF6
307.03
MFCD00010306
SWFFFUJOWAJJCH-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide
518871
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr
This item is not returnable. View return policy
140690-56-8
C9H5BrF6
307.03
MFCD00010306
SWFFFUJOWAJJCH-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide
518871
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 307.03
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2, 4-Bis(trifluoromethyl)benzyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Bis(trifluoromethyl)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'