missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Biphenylyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64321.06

Code : NEW

Additional Details : CAS Number : 2562-77-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15421479

  • £498.20 / 5g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Biphenylyl)benzimidazole
2562-77-8
270.33
MFCD01159512

Specifications

2-(4-Biphenylyl)benzimidazole
Solid
270.33
5g
2562-77-8
95%
MFCD01159512
Videos
SDS