missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Biphenylyl)-5-chlorobenzimidazole, 95%

Brand:  Alfa Aesar™ H64190.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15411289

  • £119.78 / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Biphenylyl)-5-chlorobenzimidazole
304.78
MFCD22192891

Specifications

2-(4-Biphenylyl)-5-chlorobenzimidazole
95%
MFCD22192891
Solid
304.78
1g
Videos
SDS