2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 185090050

5GR 2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 16182-15-3 Weight: 0.05500kg Transport: UN number : 1325 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 5g

Product Code. 10536451

Quantity Price
1 £ 21.0 / 5g
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
CAS 16182-15-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C9H14N2O2S
Linear Formula (CH3)3C6H2SO2NHNH2
Formula Weight 214.28
MDL Number MFCD00007585
Synonym MSH

Specification

Assay ≥96%
Physical Form Crystalline Powder
Formula Weight 214.28
Melting Point 110° to 114°C (dec.)
Quantity 5g
Color White to off-white
CAS Min % 96.0
EINECS Number 240-319-4
Linear Formula (CH3)3C6H2SO2NHNH2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 16182-15-3
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00007585
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
Molecular Formula C9H14N2O2S
Health Hazard 1 Warning
CAS Max % 100.0
Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
TSCA TSCA
Synonym MSH
Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.