missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%, Thermo Scientific™

Catalog Number p-206509
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16182-15-3
MFCD00007585
JQUBKTQDNVZHIY-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
85321
2,4,6-trimethylbenzenesulfonohydrazide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
This item is not returnable. View return policy
16182-15-3
MFCD00007585
JQUBKTQDNVZHIY-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
85321
2,4,6-trimethylbenzenesulfonohydrazide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H14N2O2S
Linear Formula (CH3)3C6H2SO2NHNH2
Quantity 25 g
Molecular Weight (g/mol) 214.28
Formula Weight 214.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trimethylbenzenesulfonyl hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'