missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzene-1-thiol, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | (CH3)3C6H2SH |
Quantity | 25 g |
Formula Weight | 152.26 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4, 6-Trimethylbenzene-1-thiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'