missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzene-1-thiol, 97%, Maybridge

Brand:  Maybridge BTB12350DA

Code : 31

Additional Details : CAS Number : 1541-10-2 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10752653

  • £15.00 / 1g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

2, 4, 6-Trimethylbenzene-1-thiol
96.5
97%
C9H12S
152.26
1541-10-2
100.0
97%
(CH3)3C6H2SH
MFCD00218475

Specifications

2, 4, 6-Trimethylbenzene-1-thiol
1g
96.5
Amber Glass Bottle
97%
(CH3)3C6H2SH
MFCD00218475
1.0240g/mL
1541-10-2
100.0
97%
C9H12S
152.26
Videos
SDS