missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triisopropylbenzoyl chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
Specifications
Melting Point | 78°C to 82°C |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 2269534 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 266.81 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,4,6-Triisopropylbenzoyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'