missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzaldehyde 97%, ACROS Organics™

Brand:  Acros Organics 384200050

Code : 29

Additional Details : CAS Number : 58551-83-0 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10742134

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2,4,6-Trifluorobenzaldehyde
96.0
97%
C7H3F3O
MFCD00061197
58551-83-0
100.0
97%
160.09

Specifications

2,4,6-Trifluorobenzaldehyde
58551-83-0
100.0
97%
C7H3F3O
160.09
Glass bottle
White to slightly grey
5g
96.0
Powder
97%
Authentic
MFCD00061197
62.0° to 66.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4,6-Trifluorobenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS