missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate. It is a versatile intermediate in the synthesis of reactive dyes.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, well ventilated area. Store cold 2-4°C.
It is used as pharmaceutical intermediate. It is a versatile intermediate in the synthesis of reactive dyes.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, well ventilated area. Store cold 2-4°C.
Specifications
Melting Point | 131°C to 132°C |
Color | White to Brown |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 744505 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 211.43 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2,4,6-Trichloropyrimidine-5-carboxaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'