missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7058411
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4136-95-2
C7H2Cl4O
243.89
MFCD00075323
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride, benzoyl chloride, 2,4,6-trichloro, mi, acmc-209jk3, ksc494s2p, 2,4,6-cl3c6h2cocl, 2,4,6-ttrichlorobenzoyl chloride, 2,4,6-trichloro benzoyl chloride, 2,4,6-trichlorobenzoic acid chloride, 2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl
This item is not returnable. View return policy
4136-95-2
C7H2Cl4O
243.89
MFCD00075323
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride, benzoyl chloride, 2,4,6-trichloro, mi, acmc-209jk3, ksc494s2p, 2,4,6-cl3c6h2cocl, 2,4,6-ttrichlorobenzoyl chloride, 2,4,6-trichloro benzoyl chloride, 2,4,6-trichlorobenzoic acid chloride, 2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl

2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.561
Boiling Point 107°C to 108°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.575
Quantity 5 g
UN Number UN3265
Beilstein 2050280
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 243.91
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichlorobenzoyl chloride
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'