missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromotoluene, 97%, Thermo Scientific™

Catalog Number p-3598256
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6320-40-7
C7H5Br3
328.83
MFCD00013527
BFRIZWKDNUHPHL-UHFFFAOYSA-N
33916
1,3,5-tribromo-2-methylbenzene
CC1=C(C=C(C=C1Br)Br)Br
This item is not returnable. View return policy
6320-40-7
C7H5Br3
328.83
MFCD00013527
BFRIZWKDNUHPHL-UHFFFAOYSA-N
33916
1,3,5-tribromo-2-methylbenzene
CC1=C(C=C(C=C1Br)Br)Br

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 68°C to 71°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H2Br3
Quantity 5 g
Synonym 2,4,6-tribromotoluene, benzene, 1,3,5-tribromo-2-methyl, acmc-20aof7, 2,4,6,tribromotoluene, 2,4,6-tribrom-toluol, 2-methyl-1,3,5-tribromobenzene, benzene,1,3,5-tribromo-2-methyl, 2,4,6-tribromotoluene 10g, 1,3,5-tribromo-2-methyl-benzene
Formula Weight 328.83
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 2, 4, 6-Tribromotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Tribromotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'