missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,6-Tribromoanisole, 99%

Brand:  Alfa Aesar™ H55684.06

Code : NEW

Additional Details : CAS Number : 607-99-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402008

  • £37.47 / 5g

Estimated Shipment: 24-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

607-99-8
344.83
YXTRCOAFNXQTKL-UHFFFAOYSA-N
11839
COC1=C(Br)C=C(Br)C=C1Br
C7H5Br3O
MFCD00192510
2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5
1,3,5-tribromo-2-methoxybenzene

Specifications

2,4,6-Tribromoanisole
Brown to White
5g
C7H5Br3O
2210361
YXTRCOAFNXQTKL-UHFFFAOYSA-N
1,3,5-tribromo-2-methoxybenzene
11839
Solid
297°C to 299°C
84°C to 88°C
607-99-8
MFCD00192510
2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5
COC1=C(Br)C=C(Br)C=C1Br
344.83
344.83
99%
Videos
SDS Documents
Certificates