missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoanisole, 99%, Thermo Scientific™

Catalog Number p-7052757
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
607-99-8
C7H5Br3O
344.83
MFCD00192510
YXTRCOAFNXQTKL-UHFFFAOYSA-N
2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5
11839
1,3,5-tribromo-2-methoxybenzene
COC1=C(Br)C=C(Br)C=C1Br
This item is not returnable. View return policy
607-99-8
C7H5Br3O
344.83
MFCD00192510
YXTRCOAFNXQTKL-UHFFFAOYSA-N
2,4,6-tribromoanisole, anisole, 2,4,6-tribromo, benzene, 1,3,5-tribromo-2-methoxy, unii-do7m3m4lx5, 1,3,5-tribromo-2-methoxy-benzene, methyl 2,4,6-tribromophenyl ether, do7m3m4lx5, 2,6-tribromoanisole, anisole,4,6-tribromo, acmc-209mm5
11839
1,3,5-tribromo-2-methoxybenzene
COC1=C(Br)C=C(Br)C=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 84°C to 88°C
Color Brown to White
Boiling Point 297°C to 299°C
Quantity 1 g
Beilstein 2210361
Formula Weight 344.83
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 2,4,6-Tribromoanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'