missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-202224
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
1004-38-2
MFCD00006100
JTTIOYHBNXDJOD-UHFFFAOYSA-N
2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidine
13863
CHEBI:39857
pyrimidine-2,4,6-triamine
C1=C(N=C(N=C1N)N)N
This item is not returnable. View return policy
1004-38-2
MFCD00006100
JTTIOYHBNXDJOD-UHFFFAOYSA-N
2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidine
13863
CHEBI:39857
pyrimidine-2,4,6-triamine
C1=C(N=C(N=C1N)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 252°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C4H7N5
Quantity 5 g
Beilstein 24,47
Solubility Information Solubility in water: 36.5g/kg in water (20°C). Other solubilities: 22.7g/kg in methanol (20°C),4.5g/kg in ethanol (20°C)
Molecular Weight (g/mol) 125.13
Formula Weight 125.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Triaminopyrimidine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Triaminopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'