missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylnitrosobenzene, 99%, ACROS Organics™

Brand:  Acros Organics 207980010

Code : 29

Additional Details : CAS Number : 24973-59-9 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10742441

  • £120.50 / 1g


Estimated Shipment
13-02-2018
Add to basket

Specifications

Chemical Identifiers

2, 4, 6-Tri-tert-butylnitrosobenzene
24973-59-9
100.0
99%
{(CH3)3C}3C6H2NO
MFCD00008817
99%
98.5
≥98.5%
C18H29NO
275.42
BNB

Specifications

2, 4, 6-Tri-tert-butylnitrosobenzene
24973-59-9
100.0
Glass bottle
≥98.5%
C18H29NO
275.42
BNB
Green
99%
98.5
Authentic
Powder
99%
{(CH3)3C}3C6H2NO
MFCD00008817
170° to 173°C
1g
Videos
SDS