missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide, 97%

Brand:  Alfa Aesar™ H63514.14

Code : NEW

Additional Details : CAS Number : 256412-89-2 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15405625

  • £162.50 / 25g


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Description

Description

Applications
An aryloxyphenoxypropionate (AOPP) post-emergence plant control that shows an exclusive whole plant safety to rice with a high control efficacy to annual grass weeds, especially barnyardgrass.

Solubility
Soluble in methanol, chloroform and dimethyl sulfoxide.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide
440.86
UN3077
256412-89-2
MFCD11111427

Specifications

2-[4-(6-Chlorobenzo[d]oxazol-2-yloxy)phenoxy]-N-(2-fluorophenyl)-N-methylpropionamide
25g
97%
MFCD11111427
77° to 81°C
256412-89-2
440.86
UN3077
Videos
SDS