missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.
Solubility
Miscible with most organic solvents.
Notes
Moisture sensitive. Incompatible with acids, bases and strong oxidizing agents.
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.
Solubility
Miscible with most organic solvents.
Notes
Moisture sensitive. Incompatible with acids, bases and strong oxidizing agents.
Specifications
Boiling Point | 110°C (10mmHg) |
Melting Point | -43°C |
Quantity | 50 g |
Molecular Formula | C12H24O4Si4 |
Beilstein | 1807272 |
Flash Point | 112°C (233°F) |
Synonym | 2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl, 1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane, 2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane, tetravinyltetramethylcyclotetrasiloxane, s-tetramethyltetravinylcyclotetrasiloxane, unii-3nt606t1vk, cyclotetrasiloxane, tetraethenyltetramethyl |
Solubility Information | Miscible with most organic solvents. |
InChI Key | VMAWODUEPLAHOE-UHFFFAOYSA-N |
SMILES | C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'