missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetrachloroquinazoline, 97%, Thermo Scientific™

Catalog Number p-7055398
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
54185-42-1
C8H2Cl4N2
267.918
MFCD03844695
GYUFSVVBAINEBI-UHFFFAOYSA-N
quinazoline, 2,4,6,8-tetrachloro
12703363
2,4,6,8-tetrachloroquinazoline
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl
This item is not returnable. View return policy
54185-42-1
C8H2Cl4N2
267.918
MFCD03844695
GYUFSVVBAINEBI-UHFFFAOYSA-N
quinazoline, 2,4,6,8-tetrachloro
12703363
2,4,6,8-tetrachloroquinazoline
C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4,6,8-Tetrachloroquinazoline is employed as a intermediate for pharmaceutical and organic synthesis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 267.93
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6,8-Tetrachloroquinazoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'