missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,5-Trifluorotoluene, 98%

Brand:  Alfa Aesar™ H63190.06

Code : NEW

Additional Details : CAS Number : 887267-34-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459748

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2,4,5-Trifluorotoluene
146.11
1,2,4-Trifluoro-5-methylbenzene
887267-34-7
MFCD08458011

Specifications

2,4,5-Trifluorotoluene
Colorless to pale yellow
Liquid
146.11
1,2,4-Trifluoro-5-methylbenzene
887267-34-7
5g
98%
MFCD08458011
Videos
SDS