2,4,5-Trifluorobenzoyl chloride, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 309330010

1GR 2,4,5-Trifluorobenzoyl chloride, 99%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 88419-56-1 Weight: 0.05100kg Transport: UN number : 3265 Chem class : 1a Pack group : III


Packaging Glass bottle
Quantity 1g

Product Code. 10617451

Quantity Price
1 £ 20.4 / 1g
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
CAS 88419-56-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H2ClF3O
Linear Formula F3C6H2COCl
Formula Weight 194.54
MDL Number MFCD00061204

Specification

Assay ≥98.5%
Physical Form Liquid
Formula Weight 194.54
Quantity 1g
Color Clear colorless
CAS Min % 98.5
Linear Formula F3C6H2COCl
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 88419-56-1
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Assay Percent Range 99%
MDL Number MFCD00061204
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Molecular Formula C7H2ClF3O
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Density 1.5200g/mL
Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.