missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 384190010

Code : 29

Additional Details : CAS Number : 165047-24-5 Weight : 0.05100kg Transport : UN number : 1989 Chem class : 3 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10765905

  • £7.80 / 1g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

2, 4, 5-Trifluorobenzaldehyde
96.0
97%
C7H3F3O
160.09
165047-24-5
100.0
97%
F3C6H3CHO
MFCD00061196

Specifications

2, 4, 5-Trifluorobenzaldehyde
58°C
Clear colorless to light yellow
165047-24-5
100.0
Glass bottle
97%
C7H3F3O
160.09
1.4080g/mL
168.0°C
1g
96.0
Authentic
Liquid
97%
F3C6H3CHO
MFCD00061196
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4,5-Trifluorobenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS