missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 384190050

Code : 29

Additional Details : CAS Number : 165047-24-5 Weight : 0.05500kg Transport : UN number : 1989 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10567634

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2, 4, 5-Trifluorobenzaldehyde
96.0
≥96.0%
C7H3F3O
160.09
165047-24-5
100.0
97%
F3C6H3CHO
MFCD00061196

Specifications

2, 4, 5-Trifluorobenzaldehyde
Clear colorless to light yellow
165047-24-5
100.0
Glass bottle
≥96.0%
C7H3F3O
160.09
1.408g/mL
5g
96.0
Authentic
Liquid
97%
F3C6H3CHO
MFCD00061196
Videos
SDS