missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-2-pentene, 98%, ACROS Organics™

Brand:  Acros Organics 162631000

Code : 29

Additional Details : CAS Number : 107-40-4 Weight : 0.12200kg Transport : UN number : 2050 Chem class : 3 Pack group : II

Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10782581

  • £208.40 / 100mL


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

2, 4, 4-Trimethyl-2-pentene
107-40-4
100.0
98%
(CH3)3CCH=C(CH3)2
MFCD00008902
98%
97.5
98%
C8H16
112.21
β-Diisobutylene, β-Diisobutylene

Specifications

2, 4, 4-Trimethyl-2-pentene
2°C
104.0°C
− 106.0°C
98%
97.5
Authentic
1.4150 to 1.4170
98%
C8H16
112.21
01, III, 848
0.7200g/mL
0.72
Clear colorless to very slightly yellow
100mL
107-40-4
100.0
Glass bottle
Liquid
98%
(CH3)3CCH=C(CH3)2
MFCD00008902
β-Diisobutylene, β-Diisobutylene
Videos
SDS