missing translation for 'onlineSavingsMsg'
Learn More

2,4,4'-Trimethoxybenzophenone, 99%, Thermo Scientific™

Catalog Number 11313157
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
4038-15-7
C16H16O4
272.3
MFCD00100089
VFTDVICZBGDMMB-UHFFFAOYSA-N
2,4,4'-trimethoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl methanone, acmc-20aofo, maybridge1_005563, 2,4-dimethoxy-4'-methoxybenzophenone, 2, 4-dimethoxy-4'-methoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl ketone, 2,4-dimethoxyphenyl-4-methoxyphenyl methanone, methanone, 2,4-dimethoxyphenyl 4-methoxyphenyl, 2,4-dimethoxy-phenyl-4-methoxy-phenyl-methanone
254174
(2,4-dimethoxyphenyl)-(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)OC
This item is not returnable. View return policy
4038-15-7
C16H16O4
272.3
MFCD00100089
VFTDVICZBGDMMB-UHFFFAOYSA-N
2,4,4'-trimethoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl methanone, acmc-20aofo, maybridge1_005563, 2,4-dimethoxy-4'-methoxybenzophenone, 2, 4-dimethoxy-4'-methoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl ketone, 2,4-dimethoxyphenyl-4-methoxyphenyl methanone, methanone, 2,4-dimethoxyphenyl 4-methoxyphenyl, 2,4-dimethoxy-phenyl-4-methoxy-phenyl-methanone
254174
(2,4-dimethoxyphenyl)-(4-methoxyphenyl)methanone
COC1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 71°C to 73°C
Quantity 10 g
Beilstein 1991889
Formula Weight 272.3
Percent Purity 99%
Chemical Name or Material 2,4,4'-Trimethoxybenzophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'