missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%, Alfa Aesar™

Brand:  Alfa Aesar H64084.06

Code : NEW

Additional Details : CAS Number : 52516-30-0 Weight : 0.00500kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15401129

  • £57.56 / 5g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

52516-30-0
189.181
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
104223
C1=CC(=CC(=C1)C(F)(F)F)CCN
C9H10F3N
MFCD00040756
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
2-[3-(trifluoromethyl)phenyl]ethanamine

Specifications

2-[3-(Trifluoromethyl)phenyl]ethylamine
1.4645
113°C (22mmHg)
5g
C9H10F3N
UN2735
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
C1=CC(=CC(=C1)C(F)(F)F)CCN
189.181
189.18
98%
1.18 g/mL
Light sensitive
Colorless to Yellow
52516-30-0
MFCD00040756
4398117
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]ethanamine
104223
Liquid
Videos
SDS Documents
Certificates