missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Thienyl)ethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3758810
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13781-67-4
C6H8OS
128.19
MFCD00009766
YYPNNBPPDFTQFX-UHFFFAOYSA-N
2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415
83731
OCCC1=CSC=C1
This item is not returnable. View return policy
13781-67-4
C6H8OS
128.19
MFCD00009766
YYPNNBPPDFTQFX-UHFFFAOYSA-N
2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415
83731
OCCC1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Density 1.1400g/mL
Boiling Point 110.0°C to 111.0°C (14.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5510 to 1.5530
Quantity 5 g
Beilstein 17, IV, 1277
Specific Gravity 1.14
IUPAC Name 2-(thiophen-3-yl)ethan-1-ol
Formula Weight 128.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(3-Thienyl)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'