missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Pyridyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62079.03

Code : NEW

Additional Details : CAS Number : 700-93-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408668

  • £72.50 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Pyridyl)thiazolidine
166.24
3-(2-Thiazolidinyl)pyridine
700-93-6
MFCD01691463

Specifications

2-(3-Pyridyl)thiazolidine
700-93-6
97%
MFCD01691463
1g
Solid
166.24
3-(2-Thiazolidinyl)pyridine
Videos
SDS