missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Pyridyl)ethan-1-ol, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 123.15 |
Percent Purity | 97% |
Chemical Name or Material | 2-(3-Pyridyl)ethan-1-ol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'