missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2',3'-O-Isopropylideneadenosine, 98%

Catalog Number p-151453
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
362-75-4
C13H17N5O4
307.31
MFCD00005756
LCCLUOXEZAHUNS-AUWRGFAENA-N
2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
2723654
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12
This item is not returnable. View return policy
362-75-4
C13H17N5O4
307.31
MFCD00005756
LCCLUOXEZAHUNS-AUWRGFAENA-N
2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
2723654
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.75 to 0.81 (A250/A260),0.13 to 0.17 (A280/A260),0.01 (A290/A260) max
CAS Min % 97.5
CAS Max % 100.0
Melting Point 221.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Spectrophotometry 96% min. (lambda max. 259nm,pH 7) molar absorptivity 15400+/-4%,on dry substance
Formula Weight 307.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2', 3'-O-Isopropylideneadenosine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'