missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Methylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64264.03

Code : NEW

Additional Details : CAS Number : 6528-83-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401379

  • £141.00 / 1g

Estimated Shipment: 25-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6528-83-2
208.264
USLJYASOSQJYGN-UHFFFAOYSA-N
2879476
CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2
C14H12N2
MFCD01220491
2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-3-methylphenyl-benzimidazole
2-(3-methylphenyl)-1H-benzimidazole

Specifications

2-(3-Methylphenyl)benzimidazole
1g
C14H12N2
2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-3-methylphenyl-benzimidazole
CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2
208.264
208.26
95%
White
6528-83-2
MFCD01220491
USLJYASOSQJYGN-UHFFFAOYSA-N
2-(3-methylphenyl)-1H-benzimidazole
2879476
Solid
Videos
SDS Documents
Certificates