missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Methylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64264.06

Code : NEW

Additional Details : CAS Number : 6528-83-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411379

  • £637.50 / 5g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Methylphenyl)benzimidazole
208.26
2-(m-Tolyl)benzimidazole
6528-83-2
MFCD01220491

Specifications

2-(3-Methylphenyl)benzimidazole
6528-83-2
95%
MFCD01220491
2-(m-Tolyl)benzimidazole
5g
Solid
208.26
White
Videos
SDS