missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Methyl-2-thienyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62643.06

Code : NEW

Additional Details : CAS Number : 318466-03-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15447676

  • £398.56 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Methyl-2-thienyl)thiazolidine-4-carboxylic acid
318466-03-4
229.32
MFCD00106201

Specifications

2-(3-Methyl-2-thienyl)thiazolidine-4-carboxylic acid
318466-03-4
97%
MFCD00106201
5g
Solid
229.32
Videos
SDS