missing translation for 'onlineSavingsMsg'
Learn More

2,3-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-3778922
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
583-61-9
MFCD00009605
HPYNZHMRTTWQTB-UHFFFAOYSA-N
2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
11420
2,3-dimethylpyridine
CC1=C(N=CC=C1)C
This item is not returnable. View return policy
583-61-9
MFCD00009605
HPYNZHMRTTWQTB-UHFFFAOYSA-N
2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
11420
2,3-dimethylpyridine
CC1=C(N=CC=C1)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -17.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 161.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 0.7% max. 2-Ethylpiridine (GC)
Packaging Glass bottle
Molecular Formula C7H9N
Refractive Index 1.5062 to 1.5082
Quantity 25 mL
Beilstein 20, 243
Specific Gravity 0.95
Solubility Information Solubility in water: 95g/L (26°C). Other solubilities: soluble in methanol and ether,miscible with dmf and thf
Molecular Weight (g/mol) 107.16
Formula Weight 107.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2, 3-Lutidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3-Lutidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'