missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62531.06

Code : NEW

Additional Details : CAS Number : 80457-74-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487536

  • £398.56 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid
80457-74-5
225.26
MFCD00179286

Specifications

2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid
80457-74-5
97%
MFCD00179286
5g
Solid
225.26
Videos
SDS