missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Fluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64628.06

Code : NEW

Additional Details : CAS Number : 324-15-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15491909

  • £314.82 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Fluorophenyl)benzimidazole
324-15-2
212.23
MFCD01564628

Specifications

2-(3-Fluorophenyl)benzimidazole
324-15-2
95%
MFCD01564628
5g
Solid
212.23
Videos
SDS